accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 175204-81-6

CH3NNOCl

CAS RN 175204-81-6

IUPAC Name: 4-chloro-1-methyl-1H-pyrazole-3-carbaldehyde
Synonyms: 4-chloro-1-methylpyrazole-3-carbaldehyde
Molecular Weight (g/mol): 144.56
Molecular Formula: C5H5ClN2O
InChi Key: MLGDASSMRFJUMZ-UHFFFAOYSA-N
SMILES: CN1C=C(Cl)C(C=O)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results