accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 175204-21-4

IUPAC Name: methyl 2,1,3-benzothiadiazole-5-carboxylate
Synonyms: methyl 2,1,3-benzothiadiazole-5-carboxylate
Molecular Weight (g/mol): 194.21
Molecular Formula: C8H6N2O2S
InChi Key: LRMFSMKHXKDUDG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=NSN=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results