accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OFFFONHOOH

CAS RN 175203-86-8

IUPAC Name: 4-oxo-6-(trifluoromethoxy)-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: 4-oxo-6-(trifluoromethoxy)-1H-quinoline-3-carboxylic acid
Molecular Weight (g/mol): 273.17
Molecular Formula: C11H6F3NO4
InChi Key: GSUDZUNGTZIKPY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC2=CC=C(OC(F)(F)F)C=C2C1=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results