accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3SNO

CAS RN 175202-78-5

IUPAC Name: 4-(4-tert-butyl-1,3-thiazol-2-yl)benzaldehyde
Synonyms: 4-(4-tert-butyl-1,3-thiazol-2-yl)benzaldehyde
Molecular Weight (g/mol): 245.34
Molecular Formula: C14H15NOS
InChi Key: IIKIVYFYAIKVBC-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CSC(=N1)C1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results