accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNNH3CCH3NHNH2N

CAS RN 175202-00-3

IUPAC Name: 6-hydrazinyl-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
Synonyms: 6-hydrazinyl-1,3,4-trimethylpyrazolo[3,4-b]pyridine
Molecular Weight (g/mol): 191.24
Molecular Formula: C9H13N5
InChi Key: NDFXPLOECYNSHX-UHFFFAOYSA-N
SMILES: CN1N=C(C)C2=C(C)C=C(NN)N=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results