accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OFFFBr

CAS RN 17515-77-4

IUPAC Name: 2-(bromomethyl)-5-(trifluoromethyl)furan
Synonyms: 2-(bromomethyl)-5-(trifluoromethyl)furan
Molecular Weight (g/mol): 229
Molecular Formula: C6H4BrF3O
InChi Key: YNHVBNGRNNVEMD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(CBr)O1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)