accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CSNNCl

CAS RN 175137-21-0

IUPAC Name: 4-chloro-7-methylthieno[3,2-d]pyrimidine
Synonyms: 4-chloro-7-methylthieno[3,2-d]pyrimidine
Molecular Weight (g/mol): 184.64
Molecular Formula: C7H5ClN2S
InChi Key: IYJDOVYAFDVIDB-UHFFFAOYSA-N
SMILES: CC1=CSC2=C(Cl)N=CN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results