accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNClOOH

CAS RN 175137-17-4

IUPAC Name: 1-(4-chlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
Synonyms: 1-(4-chlorophenyl)-5-propylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 264.71
Molecular Formula: C13H13ClN2O2
InChi Key: RTLVBOYPVGHHGC-UHFFFAOYSA-N
SMILES: CCCC1=C(C=NN1C1=CC=C(Cl)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results