accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COFCl(E)

CAS RN 175136-82-0

IUPAC Name: (3E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
Synonyms: (3E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
Molecular Weight (g/mol): 198.62
Molecular Formula: C10H8ClFO
InChi Key: VZRBDBXFAKDNDJ-AATRIKPKSA-N
SMILES: CC(=O)\C=C\C1=C(F)C=CC=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results