accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NNHFCl

CAS RN 175135-74-7

IUPAC Name: (3-chloro-4-fluorophenyl)hydrazine
Synonyms: (3-chloro-4-fluorophenyl)hydrazine
Molecular Weight (g/mol): 160.58
Molecular Formula: C6H6ClFN2
InChi Key: DUORBNTWCCJANU-UHFFFAOYSA-N
SMILES: NNC1=CC=C(F)C(Cl)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results