accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COClBrOH3CNH2O

CAS RN 175135-60-1

IUPAC Name: 3-bromo-5-chloro-2,6-dimethoxybenzamide
Synonyms: 3-bromo-5-chloro-2,6-dimethoxybenzamide
Molecular Weight (g/mol): 294.53
Molecular Formula: C9H9BrClNO3
InChi Key: GBVBCQIVBHAEAZ-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(Br)C(OC)=C1C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results