accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFHNNHO

CAS RN 175135-15-6

IUPAC Name: 3-[6-(trifluoromethyl)-1H-1,3-benzodiazol-2-yl]propan-1-ol
Synonyms: 3-[5-(trifluoromethyl)-3H-1,3-benzodiazol-2-yl]propan-1-ol
Molecular Weight (g/mol): 244.22
Molecular Formula: C11H11F3N2O
InChi Key: FDRMNTVCDKSRNL-UHFFFAOYSA-N
SMILES: OCCCC1=NC2=CC=C(C=C2N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results