accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

(S)(S)NHHNPP

CAS RN 174677-83-9

IUPAC Name: N1,N1-bis({[2-(diphenylphosphanyl)phenyl]methyl})cyclohexane-1,2-diamine
Synonyms: N1,N1-bis({[2-(diphenylphosphanyl)phenyl]methyl})cyclohexane-1,2-diamine
Molecular Weight (g/mol): 662.8
Molecular Formula: C44H44N2P2
InChi Key: GEZITKLHMKZYMH-UHFFFAOYNA-N
SMILES: NC1CCCCC1N(CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results