accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NSNHCl

CAS RN 17452-31-2

IUPAC Name: 5-(4-chlorophenyl)-1-methyl-2,3-dihydro-1H-imidazole-2-thione
Synonyms: 5-(4-chlorophenyl)-1-methyl-3H-imidazole-2-thione
Molecular Weight (g/mol): 224.71
Molecular Formula: C10H9ClN2S
InChi Key: RGYNOPLNIDBIGT-UHFFFAOYSA-N
SMILES: CN1C(=S)NC=C1C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results