accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BFFFFCH3NNH3C

CAS RN 174501-65-6

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1-butyl-3-methylimidazolium tetrafluoroborate
Molecular Weight (g/mol): 226.03
Molecular Formula: C8H15BF4N2
InChi Key: LSBXQLQATZTAPE-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)