accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 174403-69-1

CH3FOOCH3

CAS RN 174403-69-1

IUPAC Name: methyl 4-fluoro-2-methylbenzoate
Synonyms: methyl 4-fluoro-2-methylbenzoate
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: ZQFCTCYDRQFPBU-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results