accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3C(S)OHOO

CAS RN 17407-55-5

IUPAC Name: (2S)-2-hydroxy-3-methylbutanoate
Synonyms: (S)-2-hydroxy-3-methylbutyrate
Molecular Weight (g/mol): 117.13
Molecular Formula: C5H9O3
InChi Key: NGEWQZIDQIYUNV-BYPYZUCNSA-M
SMILES: CC(C)[C@H](O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results