accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17372-52-0

IUPAC Name: 4,6-diphenyl-2H-pyran-2-one
Synonyms: 4,6-diphenylpyran-2-one
Molecular Weight (g/mol): 248.28
Molecular Formula: C17H12O2
InChi Key: DDGIHXFDWLCKRZ-UHFFFAOYSA-N
SMILES: O=C1OC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results