accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17356-19-3

HOCH

CAS RN 17356-19-3

IUPAC Name: 1-ethynylcyclopentan-1-ol
Synonyms: cyclopentanol, 1-ethynyl-
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: LQMDOONLLAJAPZ-UHFFFAOYSA-N
SMILES: OC1(CCCC1)C#C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results