accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 173336-43-1

H3CCH3H2NNO

CAS RN 173336-43-1

IUPAC Name: 2-methyl-2-(morpholin-4-yl)propan-1-amine
Synonyms: 2-methyl-2-(morpholin-4-yl)propan-1-amine
Molecular Weight (g/mol): 158.25
Molecular Formula: C8H18N2O
InChi Key: GFIBMQGRHYGHAP-UHFFFAOYSA-N
SMILES: CC(C)(CN)N1CCOCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results