CAS RN 173315-54-3
CAS RN 173315-54-3
IUPAC Name:
2-chloro-4-fluoro-3-methylbenzoic acid
Synonyms:
2-chloro-4-fluoro-3-methylbenzoic acid
Molecular Weight (g/mol):
188.58
Molecular Formula:
C8H6ClFO2
InChi Key:
DBMUFFHLBXKTDM-UHFFFAOYSA-N
SMILES:
CC1=C(F)C=CC(C(O)=O)=C1Cl
1
–
1
of
1
results
1
2-Chloro-4-fluoro-3-methylbenzoic acid, 97%, Thermo Scientific Chemicals
CAS: 173315-54-3 Molecular Formula: C8H6ClFO2 Molecular Weight (g/mol): 188.58 MDL Number: MFCD11846444 InChI Key: DBMUFFHLBXKTDM-UHFFFAOYSA-N Synonym: benzoic acid, 2-chloro-4-fluoro-3-methyl,2-chloro-4-fluoro-3-methyl-benzoic acid,2-chloro-3-methyl-4-fluoro-benzoic acid,2-chloro-4-fluoro-3-methyl benzoic acid PubChem CID: 10845315 IUPAC Name: 2-chloro-4-fluoro-3-methylbenzoic acid SMILES: CC1=C(F)C=CC(C(O)=O)=C1Cl