accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1731-88-0

H3COOCH3

CAS RN 1731-88-0

IUPAC Name: methyl tridecanoate
Synonyms: methyl cocoate
Molecular Weight (g/mol): 228.38
Molecular Formula: C14H28O2
InChi Key: JNDDPBOKWCBQSM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC(=O)OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)