accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONCH3N

CAS RN 17289-19-9

IUPAC Name: methyl 1-methyl-1H-imidazole-4-carboxylate
Synonyms: methyl 1-methylimidazole-4-carboxylate
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: KZPZTVKOJSKVBV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN(C)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results