accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17284-97-8

IUPAC Name: 3-chloro-6-hydrazinylpyridazine
Synonyms: 3-chloro-6-hydrazinylpyridazine
Molecular Weight (g/mol): 144.56
Molecular Formula: C4H5ClN4
InChi Key: FXYQRYGWWZKUFV-UHFFFAOYSA-N
SMILES: NNC1=CC=C(Cl)N=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results