accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17283-16-8

CH3OOH3C

CAS RN 17283-16-8

IUPAC Name: 2-(3,4-dimethylphenyl)acetate
Synonyms: 2-(3,4-dimethylphenyl)acetate
Molecular Weight (g/mol): 163.2
Molecular Formula: C10H11O2
InChi Key: OTTPBKINJOYJGW-UHFFFAOYSA-M
SMILES: CC1=CC=C(CC([O-])=O)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results