accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3(R)OHONHOO

CAS RN 172695-33-9

IUPAC Name: (3R)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid
Synonyms: (3R)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-methylpentanoic acid
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: KFDMTAMWOIQTOB-LJQANCHMSA-N
SMILES: CC(C)[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results