accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17257-10-2

IUPAC Name: 5-(2-phenylethynyl)thiophene-2-carbaldehyde
Synonyms: 5-(2-phenylethynyl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 212.27
Molecular Formula: C13H8OS
InChi Key: YFMUACLZRVJOBK-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)C#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results