accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17249-79-5

ClSCl

CAS RN 17249-79-5

IUPAC Name: 2,3-dichlorothiophene
Synonyms: 2,3-dichlorothiophene
Molecular Weight (g/mol): 153.02
Molecular Formula: C4H2Cl2S
InChi Key: LVNVLQIXMBTMPH-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C=CS1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)