accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 171769-74-7

BrBrBrIr

CAS RN 171769-74-7

IUPAC Name: iridium(3+) hydrate tribromide
Synonyms: iridium(3+) hydrate tribromide
Molecular Weight (g/mol): 449.94
Molecular Formula: Br3H2IrO
InChi Key: KBXXFKPVKJZGJG-UHFFFAOYSA-K
SMILES: O.[Br-].[Br-].[Br-].[Ir+3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results