accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1715-30-6

CH3CH3NH2NNHNNNH2HNNCH3CH3NH2NH2HClHCl(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 1715-30-6

IUPAC Name: 1-{N''-[6-({amino[N''-(2-ethylhexyl)carbamimidamido]methylidene}amino)hexyl]carbamimidamido}-N'-(2-ethylhexyl)methanimidamide dihydrochloride
Synonyms: 1-{N''-[6-({amino[N''-(2-ethylhexyl)carbamimidamido]methylidene}amino)hexyl]carbamimidamido}-N'-(2-ethylhexyl)methanimidamide dihydrochloride
Molecular Weight (g/mol): 581.72
Molecular Formula: C26H58Cl2N10
InChi Key: BRJJFBHTDVWTCJ-UHFFFAOYNA-N
SMILES: Cl.Cl.CCCCC(CC)CN=C(N)NC(N)=NCCCCCCN=C(N)NC(N)=NCC(CC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results