accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17126-62-4

H3COONCS

CAS RN 17126-62-4

IUPAC Name: ethyl 3-isothiocyanatopropanoate
Synonyms: ethyl 3-isothiocyanatopropanoate
Molecular Weight (g/mol): 159.2
Molecular Formula: C6H9NO2S
InChi Key: UPTRONYNXNYITM-UHFFFAOYSA-N
SMILES: CCOC(=O)CCN=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results