accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17110-51-9

H3COOOCH3CH3CH3CH3CH3(Z)

CAS RN 17110-51-9

IUPAC Name: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl (9Z)-octadec-9-en-1-yl carbonate
Synonyms: cholesteryl oleyl carbonate
Molecular Weight (g/mol): 681.14
Molecular Formula: C46H80O3
InChi Key: XMPIMLRYNVGZIA-PFONDFGANA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCOC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)