accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1711-05-3

CH3OClO

CAS RN 1711-05-3

IUPAC Name: 3-methoxybenzoyl chloride
Synonyms: 3-methoxybenzoyl chloride
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: RUQIUASLAXJZIE-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)