accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 170846-74-9

OHOOHNH2Cl

CAS RN 170846-74-9

IUPAC Name: 2-amino-2-(2-chloro-5-hydroxyphenyl)acetic acid
Synonyms: chlorohydroxyphenylglycine
Molecular Weight (g/mol): 201.61
Molecular Formula: C8H8ClNO3
InChi Key: UNIDAFCQFPGYJJ-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=C(Cl)C=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results