accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17084-13-8

KFPFFFFF

CAS RN 17084-13-8

IUPAC Name: potassium hexafluoro-λ⁵-phosphanuide
Synonyms: potassium hexafluorophosphate
Molecular Weight (g/mol): 184.06
Molecular Formula: F6KP
InChi Key: YZDGRYDIGCWVND-UHFFFAOYSA-N
SMILES: [K+].F[P-](F)(F)(F)(F)F
Molecular Weight (g/mol) 
  • (13)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (4)
Physical Form 
  • (5)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (8)
  • (3)
  • (13)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (5)
  • (4)
  • (4)