CAS RN 1708-81-2
IUPAC Name:
(3Z)-hept-3-en-1-ol
Synonyms:
(3Z)-hept-3-en-1-ol
Molecular Weight (g/mol):
114.19
Molecular Formula:
C7H14O
InChi Key:
SDZQUCJFTUULJX-PLNGDYQASA-N
SMILES:
CCC\C=C/CCO
1
–
1
of
1
results
1
cis-3-Hepten-1-ol, 97%
CAS: 1708-81-2 Molecular Formula: C7H14O Molecular Weight (g/mol): 114.19 MDL Number: MFCD00274760 InChI Key: SDZQUCJFTUULJX-PLNGDYQASA-N Synonym: cis-3-hepten-1-ol,z-hept-3-en-1-ol,3-hepten-1-ol, z,3z-hept-3-en-1-ol,unii-lb4d00v63h,z-3-heptenol,cis-3-heptenol,z-3-hepten-1-ol,3z-3-hepten-1-ol # PubChem CID: 5364517 IUPAC Name: (Z)-hept-3-en-1-ol SMILES: CCC\C=C/CCO