accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 170570-79-3

H3COFOCH3F

CAS RN 170570-79-3

IUPAC Name: 1-(3,5-difluoro-4-methoxyphenyl)ethan-1-one
Synonyms: 1-(3,5-difluoro-4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 186.16
Molecular Formula: C9H8F2O2
InChi Key: OUJZNFJFMAKGMS-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(C=C1F)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results