accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HONH2

CAS RN 170564-98-4

IUPAC Name: 3-amino-3-phenylpropan-1-ol
Synonyms: 3-amino-3-phenylpropan-1-ol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: SEQXIQNPMQTBGN-UHFFFAOYNA-N
SMILES: NC(CCO)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)