accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 169896-41-7

IUPAC Name: diethylazanide
Synonyms: diethylazanide
Molecular Weight (g/mol): 72.13
Molecular Formula: C4H10N
InChi Key: UZBQIPPOMKBLAS-UHFFFAOYSA-N
SMILES: CC[N-]CC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results