accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NOClOCH3

CAS RN 169814-48-6

IUPAC Name: 1-[3-(4-chlorophenyl)-5-methyl-1,2-oxazol-4-yl]ethan-1-one
Synonyms: 1-[3-(4-chlorophenyl)-5-methyl-1,2-oxazol-4-yl]ethanone
Molecular Weight (g/mol): 235.67
Molecular Formula: C12H10ClNO2
InChi Key: XSAMMAPYCPHHKH-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)ON=C1C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results