accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

AuClClClClH

CAS RN 16961-25-4

IUPAC Name: hydrogen tetrachlorogolduide
Synonyms: chloroauric acid
Molecular Weight (g/mol): 343.81
Molecular Formula: AuCl4H5
InChi Key: QJXOQVKCBDMTLU-UHFFFAOYSA-O
SMILES: [H+].Cl.Cl.Cl.Cl.[Au-]
Quantity 
  • (4)
  • (1)
  • (2)
  • (3)
Grade 
  • (2)
  • (5)
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)