accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 169555-95-7

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(pyridin-4-yl)propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoic acid
Molecular Weight (g/mol): 388.42
Molecular Formula: C23H20N2O4
InChi Key: SCSSXJVRZMQUKA-NRFANRHFSA-N
SMILES: OC(=O)[C@H](CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results