accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3H3COONNH(S)

CAS RN 169447-86-3

IUPAC Name: tert-butyl (2S)-2-benzylpiperazine-1-carboxylate
Synonyms: tert-butyl (2S)-2-benzylpiperazine-1-carboxylate
Molecular Weight (g/mol): 276.38
Molecular Formula: C16H24N2O2
InChi Key: QKUHUJCLUFLGCI-AWEZNQCLSA-N
SMILES: CC(C)(C)OC(=O)N1CCNC[C@@H]1CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results