accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16924-00-8

FHTaHFFHFHFHHFHFKK

CAS RN 16924-00-8

IUPAC Name: dipotassium heptafluorotantalumdiuide
Synonyms: potassium heptafluorotantalate
Molecular Weight (g/mol): 399.19
Molecular Formula: F7H7K2Ta
InChi Key: BLXHHPZXEPALDX-UHFFFAOYSA-N
SMILES: F.F.F.F.F.F.F.[K+].[K+].[Ta--]
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)