accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16920-56-2

ClClClClClClKKIr

CAS RN 16920-56-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 483.11
Molecular Formula: Cl6IrK2
InChi Key: SCPDBYGTLSSBQR-UHFFFAOYSA-N
SMILES: [Cl].[Cl].[Cl].[Cl].[Cl].[Cl].[K+].[K+].[Ir]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results