accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16919-73-6

ClHPdHClClHClHHClHClKK

CAS RN 16919-73-6

IUPAC Name: dipotassium hexachloropalladiumdiuide
Synonyms: dipotassium hexachloropalladate(2-)
Molecular Weight (g/mol): 403.36
Molecular Formula: Cl6H6K2Pd
InChi Key: XISJPQLCGXNMQD-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.Cl.Cl.[K+].[K+].[Pd--]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results