accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16919-46-3

OOOOSnH3CH3CCH3CH3ClCl

CAS RN 16919-46-3

IUPAC Name: tin(4+) bis(2,4-dioxopentan-3-ide) dichloride
Synonyms: tin(4+) diacetylacetonate dichloride
Molecular Weight (g/mol): 387.83
Molecular Formula: C10H14Cl2O4Sn
InChi Key: YFSVHXIJJPZTCK-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Sn+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results