accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16905-14-9

KKPtIIIIII

CAS RN 16905-14-9

IUPAC Name: platinum(4+) dipotassium hexaiodide
Synonyms: platinum(4+) dipotassium hexaiodide
Molecular Weight (g/mol): 1034.71
Molecular Formula: I6K2Pt
InChi Key: BNJBUDJCJPWKRQ-UHFFFAOYSA-H
SMILES: [K+].[K+].[I-].[I-].[I-].[I-].[I-].[I-].[Pt+4]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results