accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 169037-23-4

OFFFOOHN

CAS RN 169037-23-4

IUPAC Name: 5-(trifluoromethoxy)-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-(trifluoromethoxy)-1H-indole-2,3-dione
Molecular Weight (g/mol): 231.13
Molecular Formula: C9H4F3NO3
InChi Key: XHAJMVPMNOBILF-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC2=C(NC(=O)C2=O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results