accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 168618-45-9

IUPAC Name: 3'-methoxy-[1,1'-biphenyl]-3-carboxylate
Synonyms: 3'-methoxy-[1,1'-biphenyl]-3-carboxylate
Molecular Weight (g/mol): 227.24
Molecular Formula: C14H11O3
InChi Key: JFPVSZXZHHOWMI-UHFFFAOYSA-M
SMILES: COC1=CC=CC(=C1)C1=CC=CC(=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results